missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ GFP Recombinant Aequorea victoria Protein, N-His Tag DFS Item

This recombinant protein is expressed from a DNA sequence encoding the Aequorea victoria GFP (AAB65663) (Ser 2-Lys 238) fused to a polyhistidine tag at the N-terminus

Manufacturer:  Life Technologies LS13105S07E50

 View more versions of this product

Catalog No. 13105S07E50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; GFP Recombinant Aequorea victoria Protein, N-His Tag; 50ug; Source: Aequorea victoria; Purity: greater than or equal to 85% by SDS-PAGE

Green Fluorescent Protein (GFP) recombinant Aequorea victoria protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein structure analysis and in cell biology research applications.&line-break;Background: The green fluorescent protein (GFP) is a protein that exhibits bright green fluorescence when exposed to blue light. The protein is in the shape of a cylinder, comprising 11 strands of β-sheet with an α-helix inside and short helical segments on the ends of the cylinder. Its ability to generate a highly visible, efficiently emitting internal fluorophore is both intrinsically fascinating and tremendously valuable. The green-fluorescent protein (GFP) of the jellyfish Aequorea victoria has long been used as a universal reporter in a broad range of heterologous living cells and organisms. GFP has become well established as a marker of gene expression and protein targeting in intact cells and organisms.
  • N-terminal Sequence Analysis: Glu 99
  • Activity: 1. Mesured by emission analysis; 2. Measure excitation and emission spectra of GFP at wavelengths of 395nM and 510nM respectively
  • Formulation: Lyophilized from sterile PBS, pH 7.5
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Manufactured by Sino Biological (China).

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
28.7kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
Green Fluorescent Protein
Assay Standard
E. coli
Lyophilized
PAGE
50μg
N-Terminal
GFP
GFP
Other Proteins
His Tag