missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GAPDH Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human GAPDH (P04406) (Met 1-Glu 335) fused to a polyhistidine tag at the N-terminus

$148.00 - $802.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Frozen
Gene Alias CDABP0047, G3PD, GAPD, mgC88685
Gene ID (Entrez) 2597
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10094H07E5 View Documents Life Technologies
LS10094H07E5
5μg Each for $148.00
 
10094H07E50 View Documents Life Technologies
LS10094H07E50
50μg Each for $802.00
 
Description

Description

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 50mM Tris, 30% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Frozen
2597
PAGE
CDABP0047, G3PD, GAPD, mgC88685

For Research Use Only. Not for use in diagnostic procedures.