Please login to your online account to display your discounted pricing

Sino Biological™ GAPDH Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human GAPDH (P04406) (Met 1-Glu 335) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS10094H07E50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10094H07E50

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 38kDa
Gene ID (Entrez) 2597
Protein Tag N-His
Format Frozen
Gene Symbol GAPDH
Protein Family Other Proteins
Expression System E. coli
Purification Method PAGE
Quantity 50μg
Structural Form Full Length/Recombinant
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias CDABP0047, G3PD, GAPD, mgC88685
Common Name Glyceraldehyde-3-Phosphate Dehydrogenase
Purity ≥90% by SDS-PAGE
Validated Application Assay Standard

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 50mM Tris, 30% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.