Please login to your online account to display your discounted pricing

Sino Biological™ GAD1 (GAD67) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human GAD1 (Q99259-1) (Met 1-Leu 594) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS12570H08B50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 12570H08B50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 68.3kDa
Structural Form Full Length/Recombinant
Common Name Glutamate Decarboxylase 1 (Brain, 67kDa)
Protein Tag His Tag
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Frozen
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Endotoxin Concentration <1 EU/μg
Gene Alias FLJ45882, GAD, SCP
Gene Symbol GAD1
Validated Application Assay Standard
Protein Family Other Proteins
Expression System Insect
Gene ID (Entrez) 2571

Glutamate Decarboxylase 1 (brain 67kDa) (GAD1), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 8.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.