missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ GAD1 (GAD67) Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human GAD1 (Q99259-1) (Met 1-Leu 594) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS12570H08B5

 View more versions of this product

Catalog No. 12570H08B5


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; GAD1 (GAD67) Recombinant Human Protein, His Tag; 5ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Glutamate Decarboxylase 1 (brain 67kDa) (GAD1), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 8.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2571
PAGE
5μg
Full Length/Recombinant
FLJ45882, GAD, SCP
GAD1
Other Proteins
His Tag
Frozen
68.3kDa
≥90% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Glutamate Decarboxylase 1 (Brain, 67kDa)
Assay Standard
Insect