Please login to your online account to display your discounted pricing

Sino Biological™ GAD1 (GAD67) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human GAD1 (Q99259-1) (Met 1-Leu 594) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS12570H08B25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 12570H08B25

  • / Each
 

Description & Specifications

Specifications

Quantity 5 x 5μg
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Purity ≥90% by SDS-PAGE
Gene ID (Entrez) 2571
Protein Family Other Proteins
Common Name Glutamate Decarboxylase 1 (Brain, 67kDa)
Molecular Weight 68.3kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Validated Application Assay Standard
Format Frozen
Gene Alias FLJ45882, GAD, SCP
Endotoxin Concentration <1 EU/μg
Purification Method PAGE
Protein Tag His Tag
Expression System Insect
Gene Symbol GAD1
Structural Form Full Length/Recombinant

Glutamate Decarboxylase 1 (brain 67kDa) (GAD1), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 8.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.