missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FTH1 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human FTH1 (P02794) (Met 1-Ser183)

$177.00 - $238.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias OK/SW-cl.84, FHC, FTH, FTHL6, PIG15, PLIF
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
13217HNAE25 Life Technologies
LS13217HNAE25
5 x 5μg Each for $177.00
13217HNAE50 Life Technologies
LS13217HNAE50
50μg Each for $238.00
Specifications

Description

Ferritin, Heavy Polypeptide 1 (FTH1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
OK/SW-cl.84, FHC, FTH, FTHL6, PIG15, PLIF
PAGE
Lyophilized
2495