Please login to your online account to display your discounted pricing

Sino Biological™ FTH1 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human FTH1 (P02794) (Met 1-Ser183)

Manufacturer: life technologies  LS13217HNAE25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 13217HNAE25

  • / Each
 

Description & Specifications

Specifications

Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Full Length/Recombinant
Protein Subtype Other Proteins
Common Name Ferritin, heavy polypeptide 1
Protein Family Other Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 2495
Molecular Weight 21.2kDa
Endotoxin Concentration <1 EU/μg
Gene Alias OK/SW-cl.84, FHC, FTH, FTHL6, PIG15, PLIF
Gene Symbol FTH1
Validated Application Assay Standard

Ferritin, Heavy Polypeptide 1 (FTH1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.