Please login to your online account to display your discounted pricing

Sino Biological™ FOLR1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human FOLR1 (NP_057937.1) (Met 1 -Met 233) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11241H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11241H08H25

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 2348
Termination C-Terminal
Expression System Mammalian
Purity ≥95% by SDS-PAGE
Purification Method PAGE
Format Lyophilized
Structural Form Full Length/Recombinant
Gene Symbol FOLR1
Validated Application Assay Standard
Endotoxin Concentration <1 EU/μg
Molecular Weight 26kDa
Quantity 5 x 5μg
Protein Tag His Tag
Common Name Folate receptor 1 (adult)
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Gene Alias FBP, FOLR, FOLR1
Protein Family Other Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Folate Receptor 1 (Adult) (FOLR1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.