Please login to your online account to display your discounted pricing

Sino Biological™ FN1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the Fragment 2 (Ser 607 - Pro 1265) of human FN1 (CAD91166) fused to a polyhistidine tag at the C-terminus

$77.00 - $172.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias FN, CIG, FNZ, MSF, ED-B, FINC, GFND, LETS, GFND2
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10314H08H25

DFS Item
 
life technologies
LS10314H08H25
5 x 5μg Each for $126.00

10314H08H50

DFS Item
 
life technologies
LS10314H08H50
50μg Each for $172.00

10314H08H5

DFS Item
 
life technologies
LS10314H08H5
5μg Each for $77.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias FN, CIG, FNZ, MSF, ED-B, FINC, GFND, LETS, GFND2

Fibronectin 1 (FN1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse APCS at 10μg/mL (100μL/well) can bind biotinylated FN1 with a linear range of 0.625 - 5μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.