Please login to your online account to display your discounted pricing

Sino Biological™ FN1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the Fragment 2 (Ser 607 - Pro 1265) of human FN1 (CAD91166) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10314H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10314H08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 73.2kDa
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant
Protein Subtype Fibronectin
Endotoxin Concentration <1 EU/μg
Gene Alias FN, CIG, FNZ, MSF, ED-B, FINC, GFND, LETS, GFND2
Common Name Fibronectin 1
Gene Symbol FN1
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Fibronectin 1 (FN1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse APCS at 10μg/mL (100μL/well) can bind biotinylated FN1 with a linear range of 0.625 - 5μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.