missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FLT4 (VEGF R3) Recombinant Human Protein (without Catalytic Activity Domain), His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ile 776) of human FLT4 (NP_002011.2) was expressed with a C-terminal polyhistidine tag

Manufacturer:  Life Technologies LS10806H08H5

 View more versions of this product

Catalog No. 10806H08H5


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; FLT4 (VEGF R3) Recombinant Human Protein (without Catalytic Activity Domain), His Tag; 5ug

Fms-Related Tyrosine Kinase 4 (FLT4), recombinant human protein is supplied as a lyopholized protein. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human VEGF-C (Mfr. No. 10542-H08H) at 10μg/mL (100μL/well) can bind human FLT4-his, the EC50 of human FLT4-his is 0.011μg/mL; 2. Measured by its ability to bind human VEGF-D (Mfr. 10557-H08H) in a functional ELISA
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5μg
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Growth Factors & Receptors