Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
missing translation for 'pleaseLogIn'

FLT1 (VEGFR1) Recombinant Human Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asn 756) of human FLT1 (P17948-1) fused to a polyhistidine tag at the C-terminus

$79.50 - $1,440.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias FLT, VEGFR1, FLT1
Gene ID (Entrez) 2321
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
10136H08H5 View Documents Life Technologies
LS10136H08H5
5μg Each for $79.50
 
10136H08HL View Documents Life Technologies
LS10136H08H250
5 x 50μg Each for $1,440.00
 
Description

Description

Fms-related tyrosine kinase 1 (FLT1/VEGFR1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to inhibit the VEGF-dependent proliferation of human umbilical vein endothelial cells (HUVEC) (Conn, G. et al., 1990 , Proc. Natl. Acad. Sci. USA 87:1323); the ED50 for this effect is typically 5-30ng/mL in the presence of 5ng/mL human VEGF165
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
2321
<1 EU/μg
FLT, VEGFR1, FLT1

For Research Use Only. Not for use in diagnostic procedures.