Please login to your online account to display your discounted pricing

Sino Biological™ FLRT3 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human FLRT3 (NP_938205.1) extracellular domain (Met 1-Pro 528) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11166H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11166H08H5

  • / Each
 

Description & Specifications

Specifications

Gene Symbol FLRT3
Molecular Weight 58kDa
Quantity 5μg
Protein Tag His Tag
Structural Form Truncated/Recombinant
Format Lyophilized
Common Name Fibronectin leucine rich transmembrane protein 3
Protein Family Cell Structure/Organelle Proteins
Validated Application Assay Standard
Gene Alias FLRT3
Endotoxin Concentration <1 EU/μg
Termination C-Terminal
Expression System Mammalian
Purity ≥90% by SDS-PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene ID (Entrez) 23767
Purification Method PAGE
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Protein Subtype FGFs (Fibroblast Growth Factors)|Fibronectin

Fibronectin Leucine Rich Transmembrane Protein 3 (FLRT3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.