missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FKBP1A (FKBP12) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human FKBP12 (NP_463460) (Met 1-Glu 108) fused to a C-terminal polyhistidine tag

$588.00 - $588.00

Specifications

Purity ≥95% by SDS-PAGE
Purification Method PAGE
Format Lyophilized
Gene Alias FKBP1A, RP11-314N13.2, FKBP-12, FKBP1, FKBP12, FKBP12C, PKC12, PKCI2, PPIASE
Gene ID (Entrez) 2280
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10268H08EL View Documents Life Technologies
LS10268H08E250
5 x 50μg Each for $588.00
 
Description

Description

FK506 binding protein 1A, 12kDa (FKBP1A), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM HEPES, 0.1 M NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥95% by SDS-PAGE
Lyophilized
2280
PAGE
FKBP1A, RP11-314N13.2, FKBP-12, FKBP1, FKBP12, FKBP12C, PKC12, PKCI2, PPIASE

For Research Use Only. Not for use in diagnostic procedures.