missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FIGF Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human VEGFD (NP_004460.1) corresponding to amino acid (Phe 93-Ser 201) fused to a C-terminal polyhistidine tag

Manufacturer:  Life Technologies LS10557H08H25

 View more versions of this product

Catalog No. 10557H08H25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; FIGF Recombinant Human Protein, His Tag; 5 x 5ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

C-Fos Induced Growth Factor (Vascular Endothelial Growth Factor D) (FIGF), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2277
PAGE
5 x 5μg
C-Terminal
VEGF (Vascular Endothelial Growth Factor)
FIGF, VEGF-D, VEGFD
FIGF
Growth Factors & Receptors
His Tag
Lyophilized
20kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
c-fos induced growth factor (vascular endothelial growth factor D)
Assay Standard
Mammalian