missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FH Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human FH (P07954-1) (Ala 45-Lys 510) fused to a polyhistidine tag at the C-terminus and an initial Met at the N-terminus

$212.00 - $960.00

Specifications

Purity >85% by SDS-PAGE
Purification Method PAGE
Format Lyophilized
Gene Alias HLRCC, LRCC, MCL, MCUL1
Gene ID (Entrez) 2271
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12115H08E25 View Documents Life Technologies
LS12115H08E25
5 x 5μg Each for $212.00
 
12115H08EL View Documents Life Technologies
LS12115H08E250
5 x 50μg Each for $960.00
 
Description

Description

Fumarate Hydratase (FH), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 10mM Tris, 5mM EDTA, 1mM DTT, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

>85% by SDS-PAGE
Lyophilized
2271
PAGE
HLRCC, LRCC, MCL, MCUL1

For Research Use Only. Not for use in diagnostic procedures.