missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FH Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human FH (P07954-1) (Ala 45-Lys 510) fused to a polyhistidine tag at the C-terminus and an initial Met at the N-terminus

Manufacturer:  Invitrogen LS12115H08E250

 View more versions of this product

Catalog No. 12115H08EL



Description

Description

Fumarate Hydratase (FH), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 10mM Tris, 5mM EDTA, 1mM DTT, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

E. coli
Assay Standard
Lyophilized
52kDa
>85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
Fumarate Hydratase
His Tag
Other Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2271
PAGE
5 x 50μg
C-Terminal
HLRCC, LRCC, MCL, MCUL1
FH
SDS
Documents

For Research Use Only. Not for use in diagnostic procedures.