Please login to your online account to display your discounted pricing

Sino Biological™ FGFR4 Recombinant Mouse Protein (without Catalytic Activity Domain), His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asp 366) of mouse FGFR4 (NP_032037.2) precursor fused to a polyhistidine tag at the C-terminus

$447.00 - $447.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Gene Alias Fgfr-4, Fgfr4, Mpk-11
Gene ID (Entrez) 14186
View More Specs
Includes:  Five vials of protein in suspension buffer

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50194M08H50

DFS Item
 
life technologies
LS50194M08H50
50μg Each for $447.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Gene Alias Fgfr-4, Fgfr4, Mpk-11
Gene ID (Entrez) 14186

Fibroblast Growth Factor Receptor 4 (FGFR4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: Measured by its ability to inhibit FGF acidic (aFGF/FGF1) dependent proliferation of Balb/c3T3 mouse embryonic fibroblasts; the ED50 for this effect is typically 55-75ng/mL
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.