Please login to your online account to display your discounted pricing

Sino Biological™ FGFR4 Recombinant Human Protein (without Catalytic Activity Domain), His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asp 369) of human FGFR4 (NP_002002.3) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10538H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10538H08H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 40kDa
Purity ≥85% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20 to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant
Protein Subtype FGFs (Fibroblast Growth Factors)|Receptor Tyrosine Kinases
Endotoxin Concentration <1 EU/μg
Gene Alias FGFR4, CD334, JTK2, mgC20292, TKF
Common Name Fibroblast growth factor receptor 4
Gene Symbol FGFR4
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Growth Factors & Receptors
Expression System Mammalian
KinaseFamily FGF Receptor Kinase Family
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 2264

Fibroblast growth factor receptor 4 (FGFR4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: Measured by its ability to inhibit FGF1-dependent proliferation of BALB/c 3T3 mouse fibroblasts, the ED50 for this effect is typically 0.1-0.5ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.