missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FGFR4 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asp 369) of human FGFR4 precursor (NP_002002.3) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer:  Life Technologies LS10538H02H25

 View more versions of this product

Catalog No. 10538H02H25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; FGFR4 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc Tag; 5 x 5ug; Source: Human; Purity: greater than or equal to 85% by SDS-PAGE

Fibroblast growth factor receptor 4 (FGFR4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to inhibit FGF1-dependent proliferation of BALB/c 3T3 mouse fibroblasts, the ED50 for this effect is typically 2-6ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2264
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
FGFs (Fibroblast Growth Factors)|Receptor Tyrosine Kinases
FGFR4, CD334, JTK2, mgC20292, TKF
FGFR4
Growth Factors & Receptors
FGF Receptor Kinase Family
Lyophilized
66kDa
5 x 5μg
Truncated/Recombinant
<1 EU/μg
Fibroblast growth factor receptor 4
Assay Standard
Mammalian
IgG1 Fc Tag