Please login to your online account to display your discounted pricing

Sino Biological™ FGFR4, Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asp 369) of human FGFR4 (NP_002002.3) precursor fused to the C-terminal polyhistidine tagged Fc region of human IgG1 at the C-terminus

$353.00 - $1,066.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Gene Alias FGFR4, CD334, JTK2, mgC20292, TKF
Gene ID (Entrez) 2264
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10538H03H50

DFS Item
 
life technologies
LS10538H03H50
50μg Each for $353.00

10538H03HL

DFS Item
 
life technologies
LS10538H03H250
5 x 50μg Each for $1,066.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Gene Alias FGFR4, CD334, JTK2, mgC20292, TKF
Gene ID (Entrez) 2264

Fibroblast Growth Factor Receptor 4 (FGFR4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: Measured by its ability to inhibit FGF acidic dependent proliferation of Balb3T3 mouse fibroblasts, the ED50 for this effect is typically 2-6ng/mL
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.