Please login to your online account to display your discounted pricing

Sino Biological™ FGFR2 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 377) of human FGFR2 (NP_000132.3) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10824H03H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10824H03HL

  • / Each
 

Description & Specifications

Specifications

Structural Form Truncated/Recombinant
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Family Growth Factors & Receptors
Protein Subtype Receptor Tyrosine Kinases|Growth Factor Receptors
Protein Tag His Tag/IgG1 Fc Tag
Validated Application Assay Standard
Gene Symbol FGFR2
Quantity 5 x 50μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Molecular Weight 67.6kDa
Gene ID (Entrez) 2263
Common Name Fibroblast growth factor receptor 2
Gene Alias BEK, BFR-1, CD332, CEK3, CFD1, ECT1, FLJ98662, JWS, K-SAM, KGFR, TK14, TK25
Expression System Mammalian
KinaseFamily FGF Receptor Kinase Family
Purity ≥85% by SDS-PAGE
Format Lyophilized
Endotoxin Concentration <1 EU/μg

Fibroblast growth factor receptor 2 (FGFR2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: Measured by its ability to inhibit FGF1-dependent proliferation of BALB/3T3 mouse fibroblasts, the ED50 for this effect is typically 3.7ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.