missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FGFR1 Recombinant Mouse Protein (without Catalytic Activity Domain), His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 376) of mouse FGFR1 isoform 1 (P16092-1) fused to a polyhistidine tag at the C-terminus

$69.50 - $1,080.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AW208770, FLG, Fgfr-1, Flt-2
Gene ID (Entrez) 14182
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50186M08H25 Life Technologies
LS50186M08H25
5 x 5μg Each for $182.00
50186M08H5 Life Technologies
LS50186M08H5
5μg Each for $69.50
50186M08HL Life Technologies
LS50186M08H250
5 x 50μg Each for $1,080.00
Specifications

Description

Fibroblast growth factor receptor 1 (FGFR1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to inhibit FGF1-dependent proliferation of BABL/c 3T3 mouse embryonic fibroblasts; the ED50 for this effect is typically 50-200ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
14182
<1 EU/μg
AW208770, FLG, Fgfr-1, Flt-2