missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FGFR1 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 285) of human FGFR1 (NP_075594.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$72.25 - $313.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias FGFR1, BFGFR, CD331, CEK, FGFBR, FLG, FLJ99988, FLT2, HBGFR, KAL2, N-SAM
Gene ID (Entrez) 2260
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10616H03H25 View Documents Life Technologies
LS10616H03H25
5 x 5μg Each for $198.00
 
10616H03H5 View Documents Life Technologies
LS10616H03H5
5μg Each for $72.25
 
10616H03H50 View Documents Life Technologies
LS10616H03H50
50μg Each for $313.00
 
Description

Description

Fibroblast growth factor receptor 1 (FGFR1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to inhibit FGF1-dependent proliferation of BALB/c 3T3 mouse fibroblasts, the ED50 for this effect is typically 2-4ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
2260
<1 EU/μg
FGFR1, BFGFR, CD331, CEK, FGFBR, FLG, FLJ99988, FLT2, HBGFR, KAL2, N-SAM

For Research Use Only. Not for use in diagnostic procedures.