missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FGF10 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human FGF10 (NP_004456.1) (Gln 38-Ser 208)

$103.00 - $1,344.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Lyophilized
Gene Alias FGF10
Gene ID (Entrez) 2255
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10573HNAE25 Life Technologies
LS10573HNAE25
5 x 5μg Each for $254.00
10573HNAE5 Life Technologies
LS10573HNAE5
5μg Each for $103.00
10573HNAE50 Life Technologies
LS10573HNAE50
50μg Each for $468.00
10573HNAEL Life Technologies
LS10573HNAE250
5 x 50μg Each for $1,344.00
Specifications

Description

Fibroblast Growth Factor 10 (FGF10), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
2255
PAGE
FGF10