Please login to your online account to display your discounted pricing

Sino Biological™ FGF10 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human FGF10 (NP_004456.1) (Gln 38-Ser 208)

Manufacturer: life technologies  LS10573HNAE50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10573HNAE50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 19.6kDa
Quantity 50μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene ID (Entrez) 2255
Purity ≥90% by SDS-PAGE
Protein Family Growth Factors & Receptors
Common Name Fibroblast Growth Factor 10
Structural Form Full Length/Recombinant
Purification Method PAGE
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Format Lyophilized
Gene Alias FGF10
Validated Application Assay Standard
Protein Tag Untagged
Gene Symbol FGF10
Expression System E. coli

Fibroblast Growth Factor 10 (FGF10), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.