Please login to your online account to display your discounted pricing

Sino Biological™ FGF10 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human FGF10 (NP_004456.1) (Gln 38-Ser 208)

Manufacturer: life technologies  LS10573HNAE250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10573HNAEL

  • / Each
 

Description & Specifications

Specifications

Quantity 5 x 50μg
Structural Form Full Length/Recombinant
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity ≥90% by SDS-PAGE
Gene Alias FGF10
Protein Tag Untagged
Validated Application Assay Standard
Gene Symbol FGF10
Expression System E. coli
Protein Family Growth Factors & Receptors
Common Name Fibroblast Growth Factor 10
Molecular Weight 19.6kDa
Purification Method PAGE
Format Lyophilized
Gene ID (Entrez) 2255
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.

Fibroblast Growth Factor 10 (FGF10), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.