missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FGF10 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human FGF10 (NP_004456.1) (Gln 38-Ser 208)

Manufacturer:  Life Technologies LS10573HNAE25

 View more versions of this product

Catalog No. 10573HNAE25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; FGF10 Recombinant Human Protein; 5 x 5ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Fibroblast Growth Factor 10 (FGF10), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2255
PAGE
5 x 5μg
Full Length/Recombinant
Fibroblast Growth Factor 10
Assay Standard
E. coli
Lyophilized
19.6kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
FGF10
FGF10
Growth Factors & Receptors
Untagged