Please login to your online account to display your discounted pricing

Sino Biological™ FGF1 (aFGF) Recombinant Mouse Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form (Phe 16-Asp 155) of mouse FGF1 (NP_034327.1)

$92.00 - $690.00

Specifications

Purity >98% by SDS-PAGE
Format Lyophilized
Gene Alias Dffrx, Fam, Fgf-1, Fgfa, FGF1
Gene ID (Entrez) 14164
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50179MNAE25

DFS Item
 
life technologies
LS50179MNAE25
5 x 5μg Each for $208.00

50179MNAE5

DFS Item
 
life technologies
LS50179MNAE5
5μg Each for $92.00

50179MNAE50

DFS Item
 
life technologies
LS50179MNAE50
50μg Each for $269.00

50179MNAEL

DFS Item
 
life technologies
LS50179MNAE250
5 x 50μg Each for $690.00
Description & Specifications

Specifications

Purity >98% by SDS-PAGE
Format Lyophilized
Gene Alias Dffrx, Fam, Fgf-1, Fgfa, FGF1
Gene ID (Entrez) 14164

Fibroblast growth factor 1 (FGF1/aFGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard or in other research applications.

  • Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse fibroblasts; the ED50 for this effect is typically 0.05-0.25ng/mL in the presence of 10μg/mL heparin
  • Formulation: Lyophilized in 20mM MES buffer, pH 6.0, 200mM NaCl, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.