Please login to your online account to display your discounted pricing

Sino Biological™ FETUB Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human FETUB (NP_055190.2) (Met 1-Pro 382) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11834H08H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11834H08HL

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 41.7kDa
Common Name Fetuin B
Gene Alias 16G2, Gugu, IRL685
Gene ID (Entrez) 26998
Format Lyophilized
Protein Family Kinases & Inhibitors/Proteases, Peptidases & Inhibitors
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Quantity 5 x 50μg
Termination C-Terminal
Endotoxin Concentration <1 EU/μg
Validated Application Assay Standard
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Tag His Tag
Purity ≥90% by SDS-PAGE
Expression System Mammalian
Structural Form Full Length/Recombinant
Purification Method PAGE
Gene Symbol FETUB

Fetuin B (FETUB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.