missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FCGR4 (CD16-2) Recombinant Mouse Protein, His.AVI Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 203) of mouse FCGR4 (NP_653142.2) fused to polyhistidine and AVI tags at the C-terminus

$121.00 - $1,338.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias 4833442P21Rik, CD16-2, FcgRIV, FcgammaRIV, FCGR3a, Fcrl3, FCGR4
Gene ID (Entrez) 246256
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50036M27H25 Life Technologies
LS50036M27H25
5 x 5μg Each for $246.00
50036M27H5 Life Technologies
LS50036M27H5
5μg Each for $121.00
50036M27H50 Life Technologies
LS50036M27H50
50μg Each for $417.00
50036M27HL Life Technologies
LS50036M27H250
5 x 50μg Each for $1,338.00
Specifications

Description

Fc gamma receptor IV (FCGR4/CD16-2), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
246256
<1 EU/μg
4833442P21Rik, CD16-2, FcgRIV, FcgammaRIV, FCGR3a, Fcrl3, FCGR4