Please login to your online account to display your discounted pricing

Sino Biological™ FCGR3B (CD16b) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 200) of human FCGR3B (NP_000561.3) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11046H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11046H08H25

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 2215
Common Name Fc fragment of IgG, low affinity IIIb, receptor (CD16b)
Validated Application Assay Standard
Expression System Mammalian
Protein Family CD Proteins
Purity ≥95% by SDS-PAGE
Molecular Weight 22.2kDa
Structural Form Truncated/Recombinant/Cell Surface Protein
Quantity 5 x 5μg
Protein Tag His Tag
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Endotoxin Concentration <1 EU/μg
Protein Subtype Glycoproteins|Fc Receptors
Gene Symbol FCGR3B
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias CD16, CD16b, FCG3, FCGR3, FCGR3B

Fc gamma receptor III-b (FCGR3B/CD16b), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.