Please login to your online account to display your discounted pricing

Sino Biological™ FCGR3A (CD16a) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1- Gln 208) of human FCGR3A (P08637-1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10389H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10389H08H5

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 2214
Format Lyophilized
Expression System Mammalian
Molecular Weight 23.3kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias RP11-5K23.1, CD16, CD16A, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3
Structural Form Truncated/Recombinant/Cell Surface Protein
Endotoxin Concentration <1 EU/μg
Protein Subtype Glycoproteins|Fc Receptors
Purity ≥90% by SDS-PAGE
Common Name Fc fragment of IgG, low affinity IIIa, receptor (CD16a)
Gene Symbol FCGR3A
Quantity 5μg
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Growth Factors & Receptors/CD Proteins

Fc gamma receptor III-a (FCGR3A/CD16a), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

  • Activity: Using the Octet RED System, the affinity constant (Kd) of CD16a bound to Human IgG1 was 180nM
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.