missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FCGR2b (CD32b) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Ala 46-Pro 217) of mature human FCGR2b (NP_001002274.1) protein was expressed with a N-terminal signal peptide and a C-terminal polyhistidine tag

$404.00 - $404.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD32, CD32B, FCG2, FCGR2, FCGR2b, IGFR2
Gene ID (Entrez) 2213
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10259H08H50 View Documents Invitrogen
LS10259H08H50
50μg Each for $404.00
 
Description

Description

Fc gamma receptor II-b (FCGR2b/CD32b), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human FCGR2b at 10μg/mL (100μL/well) can bind human IgG2 with a linear range of 0.16-6.4μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
2213
<1 EU/μg
CD32, CD32B, FCG2, FCGR2, FCGR2b, IGFR2

For Research Use Only. Not for use in diagnostic procedures.