missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FCGR2b (CD32b) Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Ala 46-Pro 217) of mature human FCGR2b (NP_001002274.1) protein was expressed with a N-terminal signal peptide and a C-terminal polyhistidine tag

Manufacturer:  Life Technologies LS10259H08H50

 View more versions of this product

Catalog No. 10259H08H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; FCGR2b (CD32b) Recombinant Human Protein, His Tag; 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Fc gamma receptor II-b (FCGR2b/CD32b), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human FCGR2b at 10μg/mL (100μL/well) can bind human IgG2 with a linear range of 0.16-6.4μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2213
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins|Fc Receptors
CD32, CD32B, FCG2, FCGR2, FCGR2b, IGFR2
FCGR2b
Growth Factors & Receptors/CD Proteins
His Tag
Lyophilized
20.8kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Fc fragment of IgG, low affinity IIb, receptor (CD32)
Assay Standard
Mammalian