missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FCGR2b (CD32) Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 217) of mouse FCGR2b precursor (NP_001070657.1) fused to a polyhistidine tag at the C-terminus

$120.00 - $1,382.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AI528646, CD32, F630109E10Rik, Fcgr2, Fcgr2a, Fcr-2, Fcr-3, Ly-17, Ly-m20, Lym-1, FCGR2b
Gene ID (Entrez) 14130
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50030M08H25 Life Technologies
LS50030M08H25
5 x 5μg Each for $240.00
50030M08H50 Life Technologies
LS50030M08H50
50μg Each for $390.00
50030M08H5 Life Technologies
LS50030M08H5
5μg Each for $120.00
50030M08HL Life Technologies
LS50030M08H250
5 x 50μg Each for $1,382.00
Specifications

Description

Fc gamma receptor II-b (FCGR2b/CD32), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
14130
<1 EU/μg
AI528646, CD32, F630109E10Rik, Fcgr2, Fcgr2a, Fcr-2, Fcr-3, Ly-17, Ly-m20, Lym-1, FCGR2b