Please login to your online account to display your discounted pricing

Sino Biological™ FCGR2A (CD32a) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ile 218) of human FCGR2A precursor (AAA35827.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10374H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10374H08H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 22kDa
Format Lyophilized
Common Name Fc fragment of IgG, low affinity IIa, receptor (CD32)
Expression System Mammalian
Gene ID (Entrez) 2212
Quantity 5μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Symbol FCGR2A
Gene Alias RP11-5K23.6, CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A, FCGR2A1, IGFR2, mgC23887, mgC30032
Validated Application Assay Standard
Endotoxin Concentration <1 EU/μg
Protein Family CD Proteins
Purity ≥90% by SDS-PAGE
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Tag His Tag
Protein Subtype Glycoproteins|Fc Receptors
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Fc gamma receptor II-a (FCGR2A/CD32a), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human FCGR2A at 10μg/mL (100μL/well) can bind human IgG2 with a linear range of 12.5-200ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.