Please login to your online account to display your discounted pricing

Sino Biological™ FCGR2A (CD32a), 167 Arg/His Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ile 218) of human FCGR2A (AAA35827.1), containing the 167 Arg/His mutation, fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10374H08H150

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10374H08H1

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 22kDa
Quantity 50μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Family CD Proteins
Validated Application Assay Standard
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Tag His Tag
Expression System Mammalian
Gene ID (Entrez) 2212
Structural Form Truncated/Recombinant/Cell Surface Protein
Common Name Fc fragment of IgG, low affinity IIa, receptor (CD32)
Purity ≥90% by SDS-PAGE
Gene Symbol FCGR2A
Gene Alias RP11-5K23.6, CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A, FCGR2A1, IGFR2, mgC23887, mgC30032
Format Lyophilized
Endotoxin Concentration <1 EU/μg
Protein Subtype Glycoproteins|Fc Receptors

Fc gamma receptor II-a (FCGR2A/CD32a), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.