Please login to your online account to display your discounted pricing

Sino Biological™ FCGR2A (CD32a), 167 Arg/His Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ile 218) of human FCGR2A (AAA35827.1), containing the 167 Arg/His mutation, fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10374H08H1250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10374H08H1L

  • / Each
 

Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Structural Form Truncated/Recombinant/Cell Surface Protein
Expression System Mammalian
Protein Tag His Tag
Molecular Weight 22kDa
Gene ID (Entrez) 2212
Format Lyophilized
Common Name Fc fragment of IgG, low affinity IIa, receptor (CD32)
Validated Application Assay Standard
Protein Subtype Glycoproteins|Fc Receptors
Gene Symbol FCGR2A
Protein Family CD Proteins
Gene Alias RP11-5K23.6, CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A, FCGR2A1, IGFR2, mgC23887, mgC30032
Quantity 5 x 50μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Endotoxin Concentration <1 EU/μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Fc gamma receptor II-a (FCGR2A/CD32a), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.