missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FCGR1A (CD64) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 288) of human FCGR1A (NP_000557.1) fused to a polyhistidine tag at the C-terminus

$123.00 - $1,414.00

Specifications

Purity >85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias FCGR1A, CD64, CD64A, FCRI, FLJ18345, IGFR1
Gene ID (Entrez) 2209
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10256H08H5 View Documents Life Technologies
LS10256H08H5
5μg Each for $123.00
 
10256H08HL View Documents Life Technologies
LS10256H08H250
5 x 50μg Each for $1,414.00
 
Description

Description

Fc gamma receptor I-a (FCGR1A/CD64), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized CD64 at 10μg/mL can bind human IgG1 with a linear range of 0.39-25ng/mL; 2. Scatchard analysis showed that the affinity constant (Kd) of CD64 bound to human IgG1 was 0.059nM
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

>85% by SDS-PAGE
Lyophilized
2209
<1 EU/μg
FCGR1A, CD64, CD64A, FCRI, FLJ18345, IGFR1

For Research Use Only. Not for use in diagnostic procedures.