missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FCGR1 (CD64) Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 297) of mouse FCGR1 precursor (NP_034316.1) fused to a polyhistidine tag at the C-terminus

$234.00 - $1,430.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias FCGR1, AI323638, AV092959, CD64, FcgammaRI, IGGHAFC
Gene ID (Entrez) 14129
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50086M08H25 View Documents Life Technologies
LS50086M08H25
5 x 5μg Each for $234.00
 
50086M08H50 View Documents Life Technologies
LS50086M08H50
50μg Each for $424.00
 
50086M08HL View Documents Life Technologies
LS50086M08H250
5 x 50μg Each for $1,430.00
 
Description

Description

Fc gamma receptor I (FCGR1/CD64), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
14129
<1 EU/μg
FCGR1, AI323638, AV092959, CD64, FcgammaRI, IGGHAFC

For Research Use Only. Not for use in diagnostic procedures.