missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FCER2 Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CD23 isoform 2 (P06734-1) extracellular domain (Asp 48-Ser 321) fused to a polyhistidine tag at the N-terminus

$402.00 - $1,258.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CLEC-6, CLEC6, CLECSF8, MCL, mgC40078, MPCL
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10261H07H50 Life Technologies
LS10261H07H50
50μg Each for $402.00
10261H07HL Life Technologies
LS10261H07H250
5 x 50μg Each for $1,258.00
Specifications

Description

Fc Fragment of IgE, low affinity II receptor (FCER2/CD23), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
CLEC-6, CLEC6, CLECSF8, MCL, mgC40078, MPCL
PAGE
Lyophilized
2208