missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FAS (CD95) Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 169) of mouse FAS (NP_032013.2) fused to a polyhistidine tag at the C-terminus

$121.00 - $1,300.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Fas, AI196731, APO1, APT1, CD95, TNFR6, Tnfrsf6, lpr
Gene ID (Entrez) 14102
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50027M08H5 Life Technologies
LS50027M08H5
5μg Each for $121.00
50027M08H25 Life Technologies
LS50027M08H25
5 x 5μg Each for $245.00
50027M08HL Life Technologies
LS50027M08H250
5 x 50μg Each for $1,300.00
Specifications

Description

Fas (TNF receptor superfamily, member 6), (CD95), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
14102
<1 EU/μg
Fas, AI196731, APO1, APT1, CD95, TNFR6, Tnfrsf6, lpr