missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

FAM3D Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human FAM3D (Q96BQ1-1) (Met 1-Phe 224) fused to a polyhistidine tag at the C-terminus

$165.00 - $165.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias UNQ567/PRO1130, EF7, OIT1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11659H08H5 View Documents Invitrogen
LS11659H08H5
5μg Each for $165.00
 
Description

Description

Family with sequence similarity 3, member D (FAM3D), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
  • This protein is manufactured by Sino Biological Inc.

    Specifications

    Specifications

    ≥90% by SDS-PAGE
    <1 EU/μg
    UNQ567/PRO1130, EF7, OIT1
    PAGE
    Lyophilized
    131177

    For Research Use Only. Not for use in diagnostic procedures.