missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ FABP6 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human FABP6 (AAH22489.1) (Met 1 -Ala 128)

$367.00 - $367.00

Specifications

Purity ≥95% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias I-15P, I-BABP, I-BALB, I-BAP, ILBP, ILBP3, ILLBP
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12579HNAE50 Life Technologies
LS12579HNAE50
50μg Each for $367.00
Specifications

Description

Fatty Acid Binding Protein 6, ileal (FABP6), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥95% by SDS-PAGE
<1 EU/μg
I-15P, I-BABP, I-BALB, I-BAP, ILBP, ILBP3, ILLBP
PAGE
Lyophilized
2172