Please login to your online account to display your discounted pricing

Sino Biological™ F9 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human F9 (AAB59620.1) (Met 1-Thr 461) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11503H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11503H08H25

  • / Each
 

Description & Specifications

Specifications

Quantity 5 x 5μg
Gene Alias RP6-88D7.1, FIX, HEMB, mgC129641, mgC129642, PTC
Gene ID (Entrez) 2158
Common Name Coagulation factor IX
Expression System Mammalian
Structural Form Full Length/Recombinant
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Endotoxin Concentration <1 EU/μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Purification Method PAGE
Protein Tag His Tag
Termination C-Terminal
Protein Family Proteases, Peptidases & Inhibitors
Purity ≥90% by SDS-PAGE
Validated Application Assay Standard
Gene Symbol F9
Molecular Weight 50kDa

Coagulation factor IX (F9), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.