missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

F2 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the full length of mouse F2 (NP_034298.1) (Met 1-Gly 618) fused to a C-terminal polyhistidine tag

$246.00 - $246.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP23-176N9.4, Cf-2, Cf2, FII, thrombin
View More Specs

Includes: Five vials of protein in suspension buffer

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50344M08H5 View Documents Life Technologies
LS50344M08H5
5μg Each for $246.00
 
Description

Description

Coagulation Factor II (F2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate Boc-VPR-AMC (R&D Systems Cat. No. ES011), the specific activity is >2000pmole/min./μg
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
RP23-176N9.4, Cf-2, Cf2, FII, thrombin
PAGE
Lyophilized
14061

For Research Use Only. Not for use in diagnostic procedures.