missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ F11R (JAMA) Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 242) of human F11R (NP_058642.1) fused to the fused Fc region of human IgG1 at the C-terminus

$366.00 - $366.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias JAM, FA11R, KAT, JAM1, JAMA, JCAM, CD321, JAM-1, JAM-A, PAM-1
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10198H08H50 Life Technologies
LS10198H08H50
50μg Each for $366.00
Specifications

Description

F11 Receptor (F11R), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its ability to support the adhesion of Jurkat human leukemic T cells; when 8 x 104 cells/well are added to F11R coated plates (0.2μg/mL and 100μL/well), approximately 30-40% will adhere specifically after 60 minutes at 37°C
  • Formulation: Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
JAM, FA11R, KAT, JAM1, JAMA, JCAM, CD321, JAM-1, JAM-A, PAM-1