missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ ERBB4 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 651) of human ERBB4 precursor (NP_001036064.1) fused to the His-tagged Fc region of human IgG1 at the C-terminus

Manufacturer:  Life Technologies LS10363H03H50

 View more versions of this product

Catalog No. 10363H03H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; ERBB4 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag; 50ug; Source: Human; Purity: greater than or equal to 85% by SDS-PAGE

V-erb-a erythroblastic leukemia viral oncogene homolog 4 (ERBB4), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human ERBB3-Fc at 1μg/well can bind biotinylated recombinant human NRG1 with a linear range of 0.032-0.16μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2066
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Receptor Tyrosine Kinases|Growth Factor Receptors
HER4, mgC138404, p180erbB4, ERBB4
ERBB4
Growth Factors & Receptors
EGF Receptor (ErbB) Kinase Family
Lyophilized
97.5kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
v-erb-a erythroblastic leukemia viral oncogene homolog 4
Assay Standard
Mammalian
His Tag/IgG1 Fc Tag